Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf165 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567233
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567233 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567233 Supplier Invitrogen™ Supplier No. PA567233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GVLSNLFTHIQ PFLGAHELYF PERVMQCHLN GATVKTDVCR MKEHMEDRVN VADFRKLEWL FPETTANFDK LLIQYRGFCA YTFAATDGLL L Highest antigen sequence identity to the following orthologs - mouse 70%, rat 70%.

CFAP206 may regulate cilium motility through its role in the assembly of the axonemal radial spokes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf165
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CFAP206
Gene Accession No. Q8IYR0
Gene Alias 1700003M02Rik; AU024269; C6orf165; Cfap206; cilia and flagella associated protein 206; Cilia- and flagella-associated protein 206; dJ382I10.1; RGD1310641; RIKEN cDNA 1700003M02 gene; similar to hypothetical protein; UPF0704 protein C6orf165; UPF0704 protein C6orf165 homolog
Gene Symbols CFAP206
Host Species Rabbit
Immunogen Recombinant Human C6orf165. Recombinant protein control fragment (Product #RP-107869).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154313
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.