Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf186 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15232R

Description
METTL24 gene ontology annotations related to this gene include methyltransferase activity.Specifications
C6orf186 | |
Polyclonal | |
Unconjugated | |
METTL24 | |
9030224M15Rik; BOS_9645; C6orf186; C9H6orf186; chromosome 6 open reading frame 186; chromosome 9 open reading frame, human C6orf186; dJ71D21.2; LOC100349157; methyltransferase like 24; Methyltransferase-like protein 24; Mettl24; UPF0624 protein C6orf186; UPF0624 protein C6orf186 homolog | |
Rabbit | |
Protein A | |
RUO | |
728464 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5JXM2 | |
METTL24 | |
KLH conjugated synthetic peptide derived from human C6orf186. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction