Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf192 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989141
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9141 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9141 Supplier Bioss Supplier No. BS9530R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a protein, which has high sequence similarity to rat, xenopus and zebrafish proteins. The protein function is unknown.

Specifications

Antigen C6orf192
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC18B1
Gene Accession No. Q6NT16
Gene Alias C6orf192; dJ55C23.6; MFS-type transporter C6orf192; MFS-type transporter SLC18B1; SLC18B1; Solute carrier family 18 member B1; solute carrier family 18, subfamily B, member 1
Gene Symbols SLC18B1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C6orf192.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116843
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.