Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf201 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541968 Supplier Invitrogen™ Supplier No. PA541968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PFGMGLGNTS RSTDAPSQST GDRKTGSVGS WGAARGPSGT DTVSGQSNSG Sequence homology: Human: 100%; Yeast: 91%.

The specific function of C6orf201 is not yet known.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf201
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C6orf201
Gene Accession No. Q7Z4U5
Gene Alias C6orf201; chromosome 6 open reading frame 201; dJ1013A10.5; protein MGC87625; uncharacterized protein C6orf201
Gene Symbols C6ORF201
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C6orf201.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 404220
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.