Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf204 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983590
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3590 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3590 Supplier Bioss Supplier No. BS15238R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene was identified as a breast cancer antigen. Nothing more is known of its function at this time. Three transcript variants encoding different isoforms have been found for this gene.

Specifications

Antigen C6orf204
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CEP85L
Gene Accession No. Q5SZL2
Gene Alias bA57K17.2; C1H6orf204; C6orf204; centrosomal protein 85 like; centrosomal protein 85kDa-like; centrosomal protein 85-like; Centrosomal protein of 85 kDa-like; CEP85L; chromosome 6 open reading frame 204-like; ENSMUSG00000038594; Gm9766; LOC100351164; NY-BR-15; rCG22129-like; Serologically defined breast cancer antigen NY-BR-15
Gene Symbols CEP85L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C6orf204.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387119
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.