Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf211 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552211
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552211 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552211 Supplier Invitrogen™ Supplier No. PA552211
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LVTDLILADF LLSSELATEV HFYGKTIPWF VSDTTIHDFN WLIEQVKHSN HKWMSKCGAD WEEYIKMGKW VYHNHIFWTL PHEYCAMPQV APDLYAELQK AHLILFKGDL NYRKLTGDRK Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 73%.

ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf211
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARMT1
Gene Accession No. Q9H993
Gene Alias Acidic residue methyltransferase 1; ARMT1; C6orf211; Damage-control phosphatase ARMT1; Protein-glutamate O-methyltransferase; Sugar phosphate phosphatase ARMT1; UPF0364 protein C6orf211
Gene Symbols ARMT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C6orf211. Recombinant protein control fragment (Product #RP-102000).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79624
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.