Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf211 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598880
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598880 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598880 Supplier Invitrogen™ Supplier No. PA598880
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C6orf211 Polyclonal Antibody for Western Blot, IHC (P), ELISA

ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf211
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ARMT1
Gene Accession No. Q9H993
Gene Alias Acidic residue methyltransferase 1; ARMT1; C6orf211; Damage-control phosphatase ARMT1; Protein-glutamate O-methyltransferase; Sugar phosphate phosphatase ARMT1; UPF0364 protein C6orf211
Gene Symbols ARMT1
Host Species Rabbit
Immunogen Recombinant Human Protein-glutamate O-methyltransferase protein (2-250AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79624
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.