Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf211 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501734737 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-4737 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-4737 Supplier Proteintech Group Inc Supplier No. 251241AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ARMT1 gene ontology annotations related to this gene include protein carboxyl O-methyltransferase activity.

Specifications

Antigen C6orf211
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ARMT1
Gene Accession No. Q9H993
Gene Alias C6orf211, UPF0364 protein C6orf211
Gene Symbols ARMT1
Host Species Rabbit
Immunogen C6orf211 Fusion Protein Ag19090
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79624
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.