Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf222 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983593
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3593 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3593 Supplier Bioss Supplier No. BS15241R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C6orf222 is a protein coding gene.

Specifications

Antigen C6orf222
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C6orf222
Gene Accession No. P0C671
Gene Alias BNIP5; C6orf222; chromosome 6 open reading frame 222; Protein BNIP5; uncharacterized protein C6orf222
Gene Symbols C6orf222
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C6orf222.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 389384
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.