Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf223 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583285
Description
Immunogen sequence: MMPLAEAGAL AQGGGPSATE WACILRRKTP RHKQPTLLMV RASRRSGKTS AVLKAGRQSV SGRKNSTSKD LVTLGASSLR E.
The exact function of MGC45491 remains unknown.
Specifications
C6orf223 | |
Polyclonal | |
Unconjugated | |
C6orf223 | |
C6orf223; chromosome 6 open reading frame 223; MGC45491; uncharacterized protein C6orf223 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
221416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N319 | |
C6orf223 | |
Recombinant protein corresponding to Human C6orf223. Recombinant protein control fragment (Product #RP-95015). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction