Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf223 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544379
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544379 Supplier Invitrogen™ Supplier No. PA544379
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CRPLRPLLGF RESDSAKPAS LRLLQHTPSA RRNYRIAGAR LMRSNYPPPL Sequence homology: Human: 100%.

The exact function of MGC45491 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf223
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C6orf223
Gene Accession No. Q8N319
Gene Alias C6orf223; chromosome 6 open reading frame 223; MGC45491; uncharacterized protein C6orf223
Gene Symbols C6orf223
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MGC45491.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221416
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.