Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf47 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565780
Description
Immunogen sequence: MFLRRLGGWLP RPWGRRKPMR PDPPYPEPRR VDSSSENSGS DWDSAPETME DVGHPKTKDS GALRVSG Highest antigen sequence identity to the following orthologs - mouse 76%, rat 75%.
C6orf47 is a protein coding gene.
Specifications
C6orf47 | |
Polyclonal | |
Unconjugated | |
C6orf47 | |
C6orf47; chromosome 6 open reading frame 47; D6S53E; G4; NG34; Protein G4; uncharacterized protein C6orf47 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57827 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95873 | |
C6orf47 | |
Recombinant Human C6orf47. Recombinant protein control fragment (Product #RP-106459). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction