Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf47 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560971 Supplier Invitrogen™ Supplier No. PA560971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EPSKEEPQVE QLGSKRMDSL KWDQPISSTQ ESGRLEAGGA SPKLRWDHVD SGGTRRPGVS PEGGLSVPGP GAPLEK Highest antigen sequence identity to the following orthologs: Mouse - 58%, Rat - 59%.

C6orf47 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf47
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C6orf47
Gene Accession No. O95873
Gene Alias C6orf47; chromosome 6 open reading frame 47; D6S53E; G4; NG34; Protein G4; uncharacterized protein C6orf47
Gene Symbols C6orf47
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C6orf47. Recombinant protein control fragment (Product #RP-99557).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57827
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.