Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf57 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA573014
Description
C6orf57 Polyclonal Antibody for Western Blot
Plays an essential role in the assembly of succinate dehydrogenase (SDH), an enzyme complex (also referred to as respiratory complex II) that is a component of both the tricarboxylic acid (TCA) cycle and the mitochondrial electron transport chain, and which couples the oxidation of succinate to fumarate with the reduction of ubiquinone (coenzyme Q) to ubiquinol (PubMed:24954416). Binds to the flavoprotein subunit Sdha in its FAD-bound form, blocking the generation of excess reactive oxigen species (ROS) and facilitating its assembly with the iron-sulfur protein subunit Sdhb into the SDH catalytic dimer.
Specifications
C6orf57 | |
Polyclonal | |
Unconjugated | |
SDHAF4 | |
1110058L19Rik; 1700001E18Rik; C6orf57; hypothetical protein LOC685888; LOC685888; SDH assembly factor 4; Sdh8; SDHAF4; Succinate dehydrogenase assembly factor 4, mitochondrial; succinate dehydrogenase complex assembly factor 4; UPF0369 protein C6orf57; UPF0369 protein C6orf57 homolog | |
Rabbit | |
Protein A | |
RUO | |
68002 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide | |
Q8BTE0 | |
SDHAF4 | |
A portion of amino acids 75-129 of mouse UPF0369 protein C6orf57 homolog was used as the immunogen. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction