Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf58 Antibody (1G1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP250604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250604 0.1 mL
NB395389 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP250604 Supplier Novus Biologicals Supplier No. NBP250604
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C6orf58 Monoclonal specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen C6orf58
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1G1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS with 0.02% Sodium Azide
Gene Alias chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003
Gene Symbols C6ORF58
Host Species Mouse
Immunogen GST-C6orf58 fusion protein
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 352999
Target Species Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.