Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf58 Antibody (1G1), Janelia Fluor™ 549, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250604JF549
Description
C6orf58 Monoclonal specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
C6orf58 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
C6ORF58 | |
GST-C6orf58 fusion protein | |
0.1 mL | |
352999 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1G1 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003 | |
Mouse | |
Protein A purified | |
Primary | |
Zebrafish | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction