Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    C6orf58 Antibody (1G1), mFluor Violet 610 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250604MFV610
Description
C6orf58 Monoclonal antibody specifically detects C6orf58 in Zebrafish samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| C6orf58 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 352999 | |
| Store at 4C in the dark. | |
| IgG1 | 
| Western Blot, Immunocytochemistry | |
| 1G1 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| chromosome 6 open reading frame 58, hypothetical protein LOC352999, MGC72003 | |
| GST-C6orf58 fusion protein | |
| 0.1 mL | |
| Primary | |
| Zebrafish | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            