Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556503
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556503 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556503 Supplier Invitrogen™ Supplier No. PA556503
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NSRKKQALNR LRAQLRKKKE SLADQFDFKM YIAFVFKEKK KKSALFEVSE VIPVMTNNYE ENILKGVRDS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf62
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C6orf62
Gene Accession No. Q9GZU0
Gene Alias 8030460C05Rik; BC005537; C6orf62; C78201; cDNA sequence BC005537; cf062; chromosome 6 open reading frame 62; dJ30M3.2; HBV XAg-transactivated protein 12; HBV X-transactivated gene 12 protein; HBV X-transactivated protein 12; Nbla00237; Uncharacterized protein C6orf62; uncharacterized protein C6orf62 homolog; XTP12
Gene Symbols C6orf62
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C6orf62. Recombinant protein control fragment (Product #RP-94685).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81688
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.