Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C6orf64 Monoclonal Antibody (OTI2H3), TrueMAB™, OriGene
SDP

Catalog No. 502540203
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0203 30 μL
50-167-5281 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0203 Supplier OriGene Supplier No. TA504635S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

C6orf64 Monoclonal antibody specifically detects C6orf64 in Canine, Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen C6orf64
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2H3
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SPATC1L
Gene Accession No. Q9H0A9
Gene Alias 1810063B07Rik; 4930488P03Rik; C12H6orf64; C23H6orf64; C6orf64; SAYSD1; SAYSvFN domain-containing protein 1; SAYSVFN motif domain containing 1; uncharacterized protein C6orf64 homolog; Unknown (protein for MGC:134290)
Host Species Mouse
Immunogen Full length human recombinant protein of human C21orf56 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 607689, 84221
Target Species Canine, Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.