Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C6orf70 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559705
Description
Immunogen sequence: KIMLPYWEVA LVKFKSHRFA DCAILLLTQL ETGLRNVFAT LNRCPKRLLT AESTALYTTF DQILAKHLND GKINQLPLFL GEPAMEFLWD FLNHQEGPRI RDHLSHGEIN LHEFSKETTN QLLAFSLVLL LRFVDDCL Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 75%.

Specifications
C6orf70 | |
Polyclonal | |
Unconjugated | |
ERMARD | |
2210404J11Rik; 2410011O22Rik; AI448938; C6orf70; dJ266L20.3; Endoplasmic reticulum membrane-associated RNA degradation protein; ER membrane associated RNA degradation; ER membrane-associated RNA degradation; ER membrane-associated RNA degradation protein; ERMARD; PVNH6; RGD1309374; transmembrane protein C6orf70; uncharacterized protein LOC381062 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55780 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5T6L9 | |
ERMARD | |
Recombinant protein corresponding to Human C6orf70. Recombinant protein control fragment (Product #RP-101015). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction