Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C6orf70 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS15252R

Specifications
C6orf70 | |
Polyclonal | |
Unconjugated | |
ERMARD | |
C6orf70; dJ266L20.3; Endoplasmic reticulum membrane-associated RNA degradation protein; ER membrane associated RNA degradation; ER membrane-associated RNA degradation protein; ERMARD; PVNH6; transmembrane protein C6orf70 | |
Rabbit | |
Protein A | |
RUO | |
55780 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5T6L9 | |
ERMARD | |
KLH conjugated synthetic peptide derived from human C6orf70. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction