Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf89 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144145
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144145 Supplier Invitrogen™ Supplier No. PA5144145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C6orf89 Polyclonal Antibody for IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf89
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene C6orf89
Gene Accession No. Q6UWU4
Gene Alias amfion; Bombesin receptor-activated protein C6orf89; BRAP; C6orf89; chromosome 6 open reading frame 89; UNQ177/PRO203
Gene Symbols C6orf89
Host Species Rabbit
Immunogen Recombinant Human Bombesin receptor-activated protein C6orf89 protein (120-320AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221477
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.