Learn More
Invitrogen™ C6orf89 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552946
Description
Immunogen sequence: QPFSPLAPEP VLSGAHTWRS LIHHIRLMSL PIAKKYMSEN KGVPLHGGDE DRPFPDFDPW WTNDCEQNES EPIPANCTGC AQKHLKVMLL EDAPRKFERL HPLVIKTGKP LLEEEIQHFL CQYPEATEGF SEGFFAKWWR CFPERWF Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 79%.

Specifications
C6orf89 | |
Polyclonal | |
Unconjugated | |
C6orf89 | |
amfion; Bombesin receptor-activated protein C6orf89; BRAP; C6orf89; chromosome 6 open reading frame 89; UNQ177/PRO203 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221477 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6UWU4 | |
C6orf89 | |
Recombinant protein corresponding to Human C6orf89. Recombinant protein control fragment (Product #RP-90591). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.