Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C6orf97 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555537 Supplier Invitrogen™ Supplier No. PA555537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ETINVHEMEA KASRETIMRL ASEVNREQKK AASCTEEKEK LNQDLLSAVE AKEALEREVK IFQERLLAGQ QVWDASKQEV SLLKKSSSEL EKSLKASQDA VT Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 68%.

The function of this gene and its encoded protein is not known. Several genome-wide association studies have implicated the region around this gene to be involved in breast cancer and bone mineral density, but no link to this specific gene has been found. [provided by RefSeq, May 2010].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C6orf97
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCDC170
Gene Accession No. Q8IYT3
Gene Alias bA282P11.1; C6orf97; CCDC170; cf097; coiled-coil domain containing 170; coiled-coil domain-containing protein 170; Gm221; RGD1310945
Gene Symbols CCDC170
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C6orf97. Recombinant protein control fragment (Product #RP-94725).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80129
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.