Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C7 Monoclonal Antibody (HB10G)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548998
Description
Antibody HM2425clone HB10G recognizes human complement C7 in its native form.
C7 is a component of the complement system. It participates in the formation of Membrane Attack Complex (MAC). People with C7 deficiency are prone to bacterial infection.
Specifications
| C7 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| A530026G17; C7; complement component 7; complement component C7; Gm1238 | |
| Mouse | |
| Protein G | |
| RUO | |
| 109828 | |
| 4°C | |
| Liquid |
| Inhibition Assays, Western Blot | |
| HB10G | |
| Unconjugated | |
| C7 | |
| C7 | |
| Truncated C7 protein covering the C-terminal end. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction