Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983609
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3609 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3609 Supplier Bioss Supplier No. BS15257R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Russel-Silver syndrome candidate,dermal papilla derived protein 13,hypothetical protein LOC79783.

Specifications

Antigen C7orf10
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Sugct
Gene Accession No. Q9HAC7
Gene Alias 5033411D12Rik; BOS_4406; C18H7orf10; C22orf6; C4H7orf10; C7orf10; caiB/baiF CoA-transferase family protein C7orf10 homolog; chromosome 4 open reading frame, human C7orf10; D17907; dermal papilla derived protein 13; Dermal papilla-derived protein 13; DERP13; GA3; hypothetical protein LOC619244; KIAA1186; KIAA1658; LOC100344733; LOW QUALITY PROTEIN: succinate--hydroxymethylglutarate CoA-transferase; ORF19; RGD1308114; Russel-Silver syndrome candidate; Succinate--hydroxymethylglutarate CoA-transferase; succinyl-CoA glutarate-CoA transferase; succinyl-CoA:glutarate-CoA transferase; SuccinylCoA:glutarate-CoA transferase; SUGCT; zgc:113390
Gene Symbols Sugct
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C7orf10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79783
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.