Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf25 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983612
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3612 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3612 Supplier Bioss Supplier No. BS15260R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C7orf25 is a protein coding gene.

Specifications

Antigen C7orf25
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene C7orf25
Gene Accession No. Q9BPX7
Gene Alias C7orf25; chromosome 7 open reading frame 25; UPF0415 protein; UPF0415 protein C7orf25
Gene Symbols C7orf25
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C7orf25.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79020
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.