Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C7orf26 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP321439AF488
Description
C7orf26 Polyclonal antibody specifically detects C7orf26 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
C7orf26 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C7orf26 chromosome 7 open reading frame 26 | |
Produced in rabbits immunized with E. coli-derived Human C7orf26 fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
79034 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction