Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf26 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA562565

Catalog No. PIPA562565


Only null left
Add to Cart

Description

Description

Immunogen sequence: AKEVLYHLDI YFSSQLQSAP LPIVDKGPVE LLEEFVFQVP KERSAQPKRL NSLQELQLLE IMCNYFQEQT KDSVRQIIFS SLFSPQGNKA DDSRMSLLGK LVSM Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 52%.

C7orf26 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C7orf26
Polyclonal
Unconjugated
C7orf26
C7orf26; chromosome 7 open reading frame 26; uncharacterized protein C7orf26
Rabbit
Antigen Affinity Chromatography
RUO
79034
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96N11
C7orf26
Recombinant protein corresponding to Human C7orf26.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.