Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf28A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560927 Supplier Invitrogen™ Supplier No. PA560927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EMPGNLQHYG RFLTGPLNLN DPDAKCRFPK IFVNTDDTYE ELHLIVYKAM SAAVCFMIDA SVHPTLDFCR RLDSIVGPQL TVLASDICEQ FNINKRMSGS EK Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Acts in concert with MON1A, as a guanine exchange factor (GEF) for RAB7, promotes the exchange of GDP to GTP, converting it from an inactive GDP-bound form into an active GTP-bound form (PubMed:23084991). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf28A
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CCZ1
Gene Accession No. P86791
Gene Alias AU022870; C25H7orf28A; C7orf28A; C7orf28B; ccz1; CCZ1 homolog, vacuolar protein trafficking and biogenesis associated; CCZ1 vacuolar protein trafficking and biogenesis associated; CCZ1 vacuolar protein trafficking and biogenesis associated homolog; CCZ1A; CCZ1B; CGI-43; H_DJ1163J12.2; H_NH0577018.2; hypothetical protein LOC393103; vacuolar fusion protein CCZ1 homolog; Vacuolar fusion protein CCZ1 homolog B; Vacuolar fusion protein CCZ1 homolog-like; zgc:55344
Gene Symbols CCZ1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C7orf28A. Recombinant protein control fragment (Product #RP-104382).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.