Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf30 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983615
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3615 Supplier Bioss Supplier No. BS15263R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Required for normal mitochondrial ribosome function and mitochondrial translation. May play a role in ribosome biogenesis by preventing premature association of the 28S and 39S ribosomal subunits. Interacts with mitochondrial ribosomal protein L14 (MRPL14), probably blocking formation of intersubunit bridge B8, preventing association of the 28S and 39S ribosomal subunits. Addition to isolated mitochondrial ribosomal subunits partially inhibits translation, probably by interfering with the association of the 28S and 39S ribosomal subunits and the formation of functional ribosomes. May also participate in the assembly and/or regulation of the stability of the large subunit of the mitochondrial ribosome. May function as a ribosomal silencing factor.

Specifications

Antigen C7orf30
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MALSU1
Gene Accession No. Q96EH3
Gene Alias C7orf30; MALSU1; mitochondrial assembly of ribosomal large subunit 1; Mitochondrial assembly of ribosomal large subunit protein 1; mtRsfA
Gene Symbols MALSU1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C7orf30.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 115416
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.