Learn More
Invitrogen™ C7orf38 Monoclonal Antibody (4E4)

Description
Peptide Sequence: MTPESRDTTD LSPGGTQEME GIVIVKVEEE DEEDHFQKER NKVESSPQVL SRSTTMNERA LLSSYLVAYR VAKEKMAHTA AEKIILPACM DMVRTIFDDK SADKLRTIPL SDNTISRRIC TIAKHLEAML ITRLQSGIDF AIQLDESTDI ASCPTLLVYV RYVWQDDFVE DLLCCLNLNS HITGLDLFTE LENCLLGQYK LNWKHCKGIS SDGTANMTGK HSRLTEKLLE ATHNNAVWNH CFIHREALVS KEISPSLMDV LKNAVKTVNF IKGSSLNSRL LEIFCSEIGV NHTHLLFHTE VRWLSQGKVL SRVYELRNEI YIFLVEKQSH LANIFEDDIW VTKLAYLSDI FGILNELSLK MQGKNNDIFQ YLEHILGFQK TLLLWQARLK SNRPSYYMFP TLLQHIEENI INEDCLKEIK LEILLHLTSL SQTFNYYFPE EKFESLKENI WMKDPFAFQN PESIIELNLE PEEENELLQL SSSFTLKNYY KILSLSAFWI KIKDDFPLLS RKSILLLLPF TTTYLCELGF SILTRLKTKK RNRLNSAPDM RVALSSCVPD WKELMNRQAH PSH.
Specifications
Specifications
| Antigen | C7orf38 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4E4 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FAM200A |
| Gene Accession No. | Q8TCP9 |
| Gene Alias | C7orf38; FAM200A; family with sequence similarity 200 member A; family with sequence similarity 200, member A; protein FAM200A |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.