Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553644 Supplier Invitrogen™ Supplier No. PA553644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ERALLSSYLV AYRVAKEKMA HTAAEKIILP ACMDMVRTIF DDKSADKLRT IPLSDNTISR RICTIAKHLE AMLITRLQSG IDFAIQLDES TDIASCPTLL VYVRYVWQDD FVEDLLCCLN LNSHITGLDL FTELENCLLG QYKLN Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 39%.

This gene encodes a protein of unknown function. The protein is weakly similar to transposase-like proteins in human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf38
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FAM200A
Gene Accession No. Q8TCP9
Gene Alias C7orf38; FAM200A; family with sequence similarity 200 member A; family with sequence similarity 200, member A; protein FAM200A
Gene Symbols FAM200A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C7orf38. Recombinant protein control fragment (Product #RP-90556).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221786
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.