Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf43 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544721
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544721 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544721 Supplier Invitrogen™ Supplier No. PA544721
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DLVERHQASL GRSQSFSHQQ PSRSHLMRSG SVMERRAITP PVASPVGRPL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.

Microtubule associated protein wich seems to play a role in spindle dynamics and cell division (PubMed:30715179, PubMed:30447097). Modulates YAP1 activity as transcriptional regulator (PubMed:30447097). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf43
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C7orf43
Gene Accession No. Q8WVR3
Gene Alias C7orf43; chromosome 7 open reading frame 43; MAP11; Microtubule-associated protein 11; RGD1305455; similar to hypothetical protein FLJ10925; uncharacterized protein C7orf43; uncharacterized protein C7orf43 homolog
Gene Symbols C7orf43
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C7orf43.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55262
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.