Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ C7orf43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556156
Description
Immunogen sequence: ETFRGEQSAF KAQVSTLLTL LPPPVLRCRQ FTVAGKHLTV LKVLNSSSQE EISIWDIRIL PNFNASYLPV MPDGSVLLVD NVCHQSGEVS MGSFCRLPGT SGCFPCPLNA LE Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Microtubule associated protein wich seems to play a role in spindle dynamics and cell division (PubMed:30715179, PubMed:30447097). Modulates YAP1 activity as transcriptional regulator (PubMed:30447097). [UniProt].
Specifications
C7orf43 | |
Polyclonal | |
Unconjugated | |
C7orf43 | |
C7orf43; chromosome 7 open reading frame 43; MAP11; Microtubule-associated protein 11; RGD1305455; similar to hypothetical protein FLJ10925; uncharacterized protein C7orf43; uncharacterized protein C7orf43 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55262 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVR3 | |
C7orf43 | |
Recombinant protein corresponding to Human C7orf43. Recombinant protein control fragment (Product #RP-92752). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction