Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546267
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546267 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546267 Supplier Invitrogen™ Supplier No. PA546267
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KNKKLKKASA EGPLLGPEAA PSGEGAGSKG EAVLRPGLDA EPELSPEEQR Sequence homology: Human: 100%.

C7orf50 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf50
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C7orf50
Gene Accession No. Q9BRJ6
Gene Alias C7orf50; chromosome 7 open reading frame 50; FP15621; MGC11257; Uncharacterized protein C7orf50; YCR016W
Gene Symbols C7orf50
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human C7orf50 (aa 15-64).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84310
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.