Learn More
Invitrogen™ C7orf50 Polyclonal Antibody

Description
C7orf50 Polyclonal Antibody for ELISA, Western Blot
Specifications
Specifications
| Antigen | C7orf50 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | C7orf50 |
| Gene Accession No. | Q9BRJ6 |
| Gene Alias | C7orf50; chromosome 7 open reading frame 50; FP15621; MGC11257; Uncharacterized protein C7orf50; YCR016W |
| Gene Symbols | C7orf50 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.