Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf50 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116589
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116589 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116589 Supplier Invitrogen™ Supplier No. PA5116589
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAKQKRKVPE VTEKKNKKLK KASAEGPLLG PEAAPSGEGA GSKGEAVLRP GLDAEPELSP EEQRVLERKL KKERKKEERQ RLREAGLVAQ HPPARRSGAE LALDYLCRWA QKHKNWRFQK TRQTWLLLHM YDSDKVPDEH FSTLLAYLEG LQGRARELTV QKAEALMREL DEEGSDPPLP GRAQRIRQVL QLLS.

C7orf50 gene ontology annotations related to this gene include RNA binding.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf50
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene C7orf50
Gene Accession No. Q9BRJ6
Gene Alias C7orf50; chromosome 7 open reading frame 50; FP15621; MGC11257; Uncharacterized protein C7orf50; YCR016W
Gene Symbols C7orf50
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-194 of human C7orf50 (NP_1157261).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84310
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.