Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf60 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565637
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565637 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565637 Supplier Invitrogen™ Supplier No. PA565637
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QERKLEQEKLS GVVKSVHRRL RKKYREVGDF DKIWREHCED EETLCEYAVA MKNLADNHWA KTCEGEGRIE WCCSVCREYF Q Highest antigen sequence identity to the following orthologs - mouse 99%, rat 38%.

S-adenosyl-L-methionine-binding protein that acts as an inhibitor of mTORC1 signaling via interaction with the GATOR1 and KICSTOR complexes (PubMed:29123071). Acts as a sensor of S-adenosyl-L-methionine to signal methionine sufficiency to mTORC1: in presence of methionine, binds S-adenosyl-L-methionine, leading to disrupt interaction with the GATOR1 and KICSTOR complexes and promote mTORC1 signaling (PubMed:29123071). Upon methionine starvation, S-adenosyl-L-methionine levels are reduced, thereby promoting the association with GATOR1 and KICSTOR, leading to inhibit mTORC1 signaling (PubMed:29123071). Probably also acts as a S-adenosyl-L-methionine-dependent methyltransferase (Potential). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf60
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene BMT2
Gene Accession No. Q1RMZ1
Gene Alias base methyltransferase of 25S rRNA 2 homolog; BMT2; C7orf60; Probable methyltransferase BMT2 homolog; probable methyltransferase BTM2 homolog; S-adenosylmethionine sensor upstream of mTORC1; SAMTOR; UPF0532 protein C7orf60
Gene Symbols BMT2
Host Species Rabbit
Immunogen Recombinant Human C7orf60. Recombinant protein control fragment (Product #RP-106306).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 154743
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.