Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf62 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113270
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113270 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113270 Supplier Invitrogen™ Supplier No. PA5113270
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C7orf62 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TEX47 (Testis Expressed 47) is a Protein Coding gene. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf62
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene TEX47
Gene Accession No. Q8TBZ9
Gene Alias C4H7orf62; C7orf62; chromosome 7 open reading frame 62; testis expressed 47; testis-expressed protein 47; TEX47; uncharacterized protein C7orf62
Gene Symbols TEX47
Host Species Rabbit
Immunogen Recombinant Human Testis-expressed protein 47 protein (1-253AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219557
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.