Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf62 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA544360
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544360 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544360 Supplier Thermo Scientific Supplier No. PA544360

Rabbit Polyclonal Antibody

Peptide sequence: EEKQRLHLKK FLLDRMFLVA KIQANVERKD VADYYEQMFQ SVLKHHLGEA Sequence homology: Cow: 77%; Human: 100%

C7orf62 Polyclonal antibody specifically detects C7orf62 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf62
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene TEX47
Gene Accession No. Q8TBZ9
Gene Alias C4H7orf62; C7orf62; chromosome 7 open reading frame 62; testis expressed 47; testis-expressed protein 47; TEX47; uncharacterized protein C7orf62
Gene Symbols TEX47
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human MGC26647
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219557
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.