Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf64 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983627
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3627 Supplier Bioss Supplier No. BS15279R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM48 gene ontology annotations related to this gene include RNA binding.

Specifications

Antigen C7orf64
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RBM48
Gene Accession No. Q5RL73
Gene Alias AW548102; BOS_4012; C030048B08Rik; C14H7orf64; C4H7orf64; C7orf64; HSPC304; MGC16142; RBM48; RGD1310794; RNA binding motif protein 48; RNA-binding protein 48; UPF0712 protein C7orf64; UPF0712 protein C7orf64 homolog
Gene Symbols RBM48
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C7orf64.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84060
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.