Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571292
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571292 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571292 Supplier Invitrogen™ Supplier No. PA571292
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 92%; Pig: 93%; Rat: 85%.

C7orf72 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf72
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SPATA48
Gene Accession No. A4D263
Gene Alias 4930415F15Rik; C18H7orf72; C4H7orf72; C7orf72; C9H7orf72; chromosome 7 open reading frame 72; chromosome 9 C7orf72 homolog; hypothetical LOC289778; Pms1; Post-meiotic spermatogenesis protein 1; RGD1309870; RIKEN cDNA 4930415F15 gene; Spata48; spermatogenesis associated 48; spermatogenesis-associated protein 48; uncharacterized protein C7orf72; uncharacterized protein C7orf72 homolog
Gene Symbols SPATA48
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human C7orf72.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100130988
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.