Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C7orf72 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555291 Supplier Invitrogen™ Supplier No. PA555291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ANIPGYTGKV HFTATHPANS NIPSTTPSPD SELHRVFQKE MAVDLFRHQA PLSRLVTTVR PYNPFNKKDK ETIDY Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 71%.

C7orf72 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C7orf72
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SPATA48
Gene Accession No. A4D263
Gene Alias 4930415F15Rik; C18H7orf72; C4H7orf72; C7orf72; C9H7orf72; chromosome 7 open reading frame 72; chromosome 9 C7orf72 homolog; hypothetical LOC289778; Pms1; Post-meiotic spermatogenesis protein 1; RGD1309870; RIKEN cDNA 4930415F15 gene; Spata48; spermatogenesis associated 48; spermatogenesis-associated protein 48; uncharacterized protein C7orf72; uncharacterized protein C7orf72 homolog
Gene Symbols SPATA48
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C7orf72. Recombinant protein control fragment (Product #RP-92787).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100130988
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.