Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C7orf72 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983630
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3630 Supplier Bioss Supplier No. BS15282R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C7orf72 is a protein coding gene.

Specifications

Antigen C7orf72
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SPATA48
Gene Alias 4930415F15Rik; C18H7orf72; C7orf72; chromosome 18 open reading frame, human C7orf72; chromosome 7 open reading frame 72; hypothetical LOC289778; Pms1; Post-meiotic spermatogenesis protein 1; RGD1309870; RIKEN cDNA 4930415F15 gene; Spata48; spermatogenesis associated 48; Spermatogenesis-associated protein 48; uncharacterized protein C7orf72; uncharacterized protein C7orf72 homolog
Gene Symbols SPATA48
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C7orf72.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100130988
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.