Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
C8A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody
$202.00 - $458.00
Specifications
| Antigen | C8A |
|---|---|
| Concentration | 0.5 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
FEPA5145448
|
Invitrogen™
PA5145448 |
20 μL |
Each for $202.00
|
|
|||||
FEPA5145447
![]() |
Invitrogen™
PA5145447 |
200 μL |
Each for $458.00
|
|
|||||
Description
C8A is a component of the complement system and contains three polypeptide subunits: alpha, beta and gamma. C8A, in particular, contains the alpha subunit of C8 and is a constituent of the membrane attack complex (MAC) that plays a key role in the innate and adaptive immune response by forming pores in the plasma membrane of target cells. C8A inserts into the target membrane, but does not form pores by itself. Disease associated with C8A include Type 1 C8 deficiency and immunodeficiency due to C8A deficiency.Specifications
| C8A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Rat | |
| C8a; complement component 8 alpha subunit; complement component 8 subunit alpha; complement component 8, alpha polypeptide; complement component C8 alpha chain | |
| C8A | |
| IgG | |
| Antigen affinity chromatography, Protein A | |
| C8A |
| 0.5 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| 298288 | |
| Recombinant Rat C8a protein, Gln21-His300 (Accession #D3ZWD6). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title