Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C8B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542539
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542539 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542539 Supplier Invitrogen™ Supplier No. PA542539
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: WKPGSSGPGS TGSWNSGSSG TGSTGNQNPG SPRPGSTGTW NPGSSERGSA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 100%.

C8 beta is one of the three subunits that comprise the component 8 (C8) of the complement system. C8 participates in the formation of Membrane Attack Complex that results in the lysis of cells. Patients with C8B deficiency are prone to bacteria infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C8B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene C8B
Gene Accession No. P07358
Gene Alias 4930439B20Rik; AI595927; C82; C8b; complement C8 beta chain; complement component 8 subunit beta; complement component 8, beta polypeptide; complement component 8, beta subunit; complement component C8 beta chain; complement component C8 beta subunit; MGC137114 protein; MGC163447
Gene Symbols C8B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human C8B.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 732
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.