Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C8G Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA551203

Catalog No. PA551203


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total C8G protein.

C8G is one of the three polypeptides that constitute C8, a component of the complement system. C8 participates in the formation of Membrane Attack Complex (MAC). Patients with deficiency in C8 are vulnerable to certain bacteria infection.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C8G
Polyclonal
Unconjugated
C8G
C8C; C8G; complement component 8, gamma polypeptide; complement component C8 gamma chain
Rabbit
Antigen Affinity Chromatography
RUO
733
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P07360
C8G
Full length fusion protein of human C8G.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.