Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ C8G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597524
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597524 Supplier Invitrogen™ Supplier No. PA597524
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

C8G Polyclonal Antibody for Western Blot, ELISA

C8G is one of the three polypeptides that constitute C8, a component of the complement system. C8 participates in the formation of Membrane Attack Complex (MAC). Patients with deficiency in C8 are vulnerable to certain bacteria infection.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C8G
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene C8G
Gene Accession No. P07360
Gene Alias C8C; C8G; complement component 8, gamma polypeptide; complement component C8 gamma chain
Gene Symbols C8G
Host Species Rabbit
Immunogen Recombinant Human Complement component C8 gamma chain protein (21-202AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 733
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.