Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C8orf30A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983632
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3632 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3632 Supplier Bioss Supplier No. BS15284R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hgh1 is a protein coding gene.

Specifications

Antigen C8orf30A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HGH1
Gene Accession No. Q9BTY7
Gene Alias brain protein 16; brain protein 16-like; Brp16; BRP16L; C8orf30A; C8orf30B; D15Ertd741e; FAM203A; FAM203B; family with sequence similarity 203, member A; family with sequence similarity 203, member B; Hgh1; HGH1 homolog; MNCb-5873; protein FAM203A; protein FAM203B; Protein HGH1 homolog
Gene Symbols HGH1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human C8orf30A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51236
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.